Learn More
Biomatik Corporation Mouse Stearoyl Coenzyme A Desaturase (SCD) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07459
Encompass_Preferred
Mouse Stearoyl Coenzyme A Desaturase (SCD) ELISA Kit for detecting SCD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13516. Gene ID: 20249. Target Synonyms: FADS5; SCDOS; Fatty Acid desaturase 5; Delta-9-Desaturase; Stearoyl-CoA Desaturase Opposite Strand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3836