Learn More
Biomatik Corporation Human Stem Cell Factor Receptor (SCFR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07467
Encompass_Preferred
Human Stem Cell Factor Receptor (SCFR) ELISA Kit for detecting SCFR in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10721. Gene ID: 3815. Target Synonyms: CD117; KIT; C-Kit; PBT; p145 c-Kit; C-Kit Receptor; Tyrosine-Protein Kinase; V-Kit Hardy-Zuckerman 4 Feline Sarcoma Viral Oncogene Homolog; Piebald Trait. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3840