Learn More
Biomatik Corporation Rat Steroid 5 Alpha Reductase 2 (SRD5a2) ELISA Kit, 96 tests
Catalog No. 501503842
Encompass_Preferred
Rat Steroid 5 Alpha Reductase 2 (SRD5a2) ELISA Kit for detecting SRD5a2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31214. Gene ID: 64677. Target Synonyms: 3-Oxo-5 Alpha-Steroid Delta 4-Dehydrogenase Alpha 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3842 Supplier Biomatik Corporation Supplier No. EKU07472
Rat Steroid 5 Alpha Reductase 2 (SRD5a2) ELISA Kit for detecting SRD5a2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31214. Gene ID: 64677. Target Synonyms: 3-Oxo-5 Alpha-Steroid Delta 4-Dehydrogenase Alpha 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.