Learn More
Biomatik Corporation Human Stratifin (SFN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07476
Encompass_Preferred
Human Stratifin (SFN) ELISA Kit for detecting SFN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31947. Gene ID: 2810. Target Synonyms: YWHAS; HME1; 14-3-3 Protein Sigma; Epithelial cell marker protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3843