Learn More
Biomatik Corporation Human Stress Induced Phosphoprotein 1 (STIP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07477
Encompass_Preferred
Human Stress Induced Phosphoprotein 1 (STIP1) ELISA Kit for detecting STIP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31948. Gene ID: 10963. Target Synonyms: HOP; P60; IEF-SSP-3521; STI1L; Hsp70/Hsp90-Organizing Protein; Renal carcinoma antigen NY-REN-11; Transformation-sensitive protein IEF SSP 3521. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3844