Learn More
Biomatik Corporation Human SFPQ (Splicing factor, proline- and glutamine-rich) ELISA Kit
Supplier:  Biomatik Corporation EKF60810
Encompass_Preferred
Human SFPQ (Splicing factor, proline- and glutamine-rich) ELISA Kit for detecting SFPQ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23246. Target Synonyms: 100 kDa DNA pairing protein; hPOMp100; POMP100; PSF; PTB associated splicing factor; SFPQ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8470