Learn More
Biomatik Corporation Pig Stromal Cell Derived Factor 1 (SDF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07480
Encompass_Preferred
Pig Stromal Cell Derived Factor 1 (SDF1) ELISA Kit for detecting SDF1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6EKW4. Gene ID: 494460. Target Synonyms: CXCL12; PBSF; P-BSF; SCYB12; SDF1-A; SDF1-B; SDF1A; SDF1B; TLSF-A; TLSF-B; TPAR1; Pre-B Cell Growth-Stimulating Factor; Chemokine C-X-C-Motif Ligand 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3847