Learn More
Biomatik Corporation Human Sulfatase 2 (SULF2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07493
Encompass_Preferred
Human Sulfatase 2 (SULF2) ELISA Kit for detecting SULF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8IWU5. Gene ID: 55959. Target Synonyms: HSULF-2; Extracellular sulfatase Sulf-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3850