Learn More
Biomatik Corporation Mouse Superoxide Dismutase 1, Soluble (SOD1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07500
Encompass_Preferred
Mouse Superoxide Dismutase 1 (SOD1) ELISA Kit for detecting SOD1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08228. Gene ID: 20655. Target Synonyms: ALS; ALS1; IPOA; Superoxide Dismutase 1, Soluble; Amyotrophic Lateral Sclerosis 1,Adult; Superoxide dismutase [Cu-Zn]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3852