Learn More
Biomatik Corporation Mouse Superoxide Dismutase 3, Extracellular (SOD3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07505
Encompass_Preferred
Mouse Superoxide Dismutase 3, Extracellular (SOD3) ELISA Kit for detecting SOD3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O09164. Gene ID: 20657. Target Synonyms: SOD-3; EC-SOD; Extracellular Superoxide Dismutase Cu-Zn. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3856