Learn More
Biomatik Corporation Human Suppressors of Cytokine Signaling 1 (SOCS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07513
Encompass_Preferred
Human Suppressors Of Cytokine Signaling 1 (SOCS1) ELISA Kit for detecting SOCS1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15524. Gene ID: 8651. Target Synonyms: CIS1; CISH1; JAB; SOCS-1; SSI-1; SSI1; TIP3; JAK-binding protein; STAT-induced STAT inhibitor 1; Tec-interacting protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3858