Learn More
Biomatik Corporation Mouse Suppressors of Cytokine Signaling 3 (SOCS3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07517
Encompass_Preferred
Mouse Suppressors Of Cytokine Signaling 3 (SOCS3) ELISA Kit for detecting SOCS3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35718. Gene ID: 12702. Target Synonyms: ATOD4; CIS3; Cish3; MGC71791; SOCS-3; SSI-3; SSI3; Cytokine-inducible SH2 protein 3; STAT-induced STAT inhibitor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3860