Learn More
Biomatik Corporation Pig Surfactant Associated Protein A (SPA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07523
Encompass_Preferred
Pig Pulmonary Surfactant Associated Protein A1 (SFTPA1) ELISA Kit for detecting SFTPA1 in serum, plasma, tissue homogenates, lung lavage fluid, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 3.12-200ng/ml. Sensitivity: 1.14ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49874. Gene ID: 397503. Target Synonyms: PRL; SFTPA; PSAP; PSPA; SFTP; SPA; COLEC4; PSP-A; SFTP1; SFTPA1B; SP-A1; Collectin-4; Alveolar proteinosis protein; 35 kDa pulmonary surfactant-associated protein; Surfactant Associated Protein A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3864