Learn More
Biomatik Corporation Human Surfactant Associated Protein B (SPB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07525
Encompass_Preferred
Human Surfactant Protein B (SP-B) ELISA Kit for detecting SP-B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07988. Gene ID: 6439. Target Synonyms: SFTPB; PSPB; SFTB3; SFTP3; SPB; Surfactant Associated Protein B; Pulmonary Surfactant Protein B; 18 kDa pulmonary-surfactant protein; 6 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3866