Learn More
Biomatik Corporation Rat NFKB-p65 (Nuclear Factor Kappa B p65) ELISA Kit
Supplier:  Biomatik Corporation EKE62221
Encompass_Preferred
Rat NFKB-p65 (Nuclear Factor Kappa B p65) ELISA Kit for detecting NFKB-p65 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.13-5000pg/ml. Sensitivity: 46.88pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: O88619. Target Synonyms: NFKB3; p65; RelA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8552