Learn More
Biomatik Corporation Human Surfactant Associated Protein D (SPD) ELISA Kit, 96 tests
Catalog No. 501503868
Encompass_Preferred
Human Surfactant Associated Protein D (SPD) ELISA Kit for detecting SPD in serum, plasma, tissue homogenates, cell lysates, lung lavage fluid, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 2.51ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35247. Gene ID: 6441. Target Synonyms: SFTPD; COLEC7; PSP-D; SFTP4; SP-D; Pulmonary Surfactant Protein D; Collectin-7; Lung surfactant protein D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3868 Supplier Biomatik Corporation Supplier No. EKU07529
Human Surfactant Associated Protein D (SPD) ELISA Kit for detecting SPD in serum, plasma, tissue homogenates, cell lysates, lung lavage fluid, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 2.51ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35247. Gene ID: 6441. Target Synonyms: SFTPD; COLEC7; PSP-D; SFTP4; SP-D; Pulmonary Surfactant Protein D; Collectin-7; Lung surfactant protein D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.