Learn More
Biomatik Corporation Rat PTGS2/COX-2 (Prostaglandin Endoperoxide Synthase 2) ELISA Kit
Supplier:  Biomatik Corporation EKE62250
Encompass_Preferred
Rat PTGS2/COX-2 (Prostaglandin Endoperoxide Synthase 2) ELISA Kit for detecting PTGS2, COX-2 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P35355. Target Synonyms: COX2; GRIPGHS; PGG/HS; PGHS-2; PHS-2; hCox-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8554