Learn More
Biomatik Corporation Rat TAT (Thrombin/Antithrombin Complex) ELISA Kit
Supplier:  Biomatik Corporation EKE62285
Encompass_Preferred
Rat TAT (Thrombin/Antithrombin Complex) ELISA Kit for detecting TAT in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.31-20ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P18292 (324-359aa). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8556