Learn More
Biomatik Corporation Rat ALPL (Alkaline Phosphatase, Liver/Bone/Kidney) ELISA Kit
Supplier:  Biomatik Corporation EKE62327
Encompass_Preferred
Rat ALPL (Alkaline Phosphatase, Liver/Bone/Kidney) ELISA Kit for detecting ALPL in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.63-40ng/ml. Sensitivity: 0.38ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P08289. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8559