Learn More
Biomatik Corporation Mouse Survivin (Surv) ELISA Kit, 96 tests
Catalog No. 501503872
Encompass_Preferred
Mouse Survivin (Surv) ELISA Kit for detecting Surv in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70201. Gene ID: 11799. Target Synonyms: BIRC5; API4; EPR-1; Baculoviral Inhibitor Of Apoptosis Repeat Containing 5; Apoptosis Inhibitor 4; Survivin Variant 3 Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3872 Supplier Biomatik Corporation Supplier No. EKU07535
Mouse Survivin (Surv) ELISA Kit for detecting Surv in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70201. Gene ID: 11799. Target Synonyms: BIRC5; API4; EPR-1; Baculoviral Inhibitor Of Apoptosis Repeat Containing 5; Apoptosis Inhibitor 4; Survivin Variant 3 Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.