Learn More
Biomatik Corporation Mouse Synapsin I (SYN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07536
Encompass_Preferred
Mouse Synapsin I (SYN1) ELISA Kit for detecting SYN1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88935. Gene ID: 20964. Target Synonyms: SYN1a; SYN1b; SYNI; Brain protein 4.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3873