Learn More
Biomatik Corporation Human Synaptophysin (SYP) ELISA Kit, 96 tests
Catalog No. 501503874
Encompass_Preferred
Human Synaptophysin (SYP) ELISA Kit for detecting SYP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08247. Gene ID: 6855. Target Synonyms: Major synaptic vesicle protein p38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3874 Supplier Biomatik Corporation Supplier No. EKU07539
Human Synaptophysin (SYP) ELISA Kit for detecting SYP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08247. Gene ID: 6855. Target Synonyms: Major synaptic vesicle protein p38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.