Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 10 (ADAM10) ELISA Kit
Supplier:  Biomatik Corporation EKL55221
Encompass_Preferred
Human A Disintegrin And Metalloprotease 10 (ADAM10) ELISA Kit for detecting ADAM10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 24.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: O14672. Gene ID: 102. Target Synonyms: CD156-C; CD156c; MADM; Kuz; CDw156; Kuzbanian protein homolog; Mammalian disintegrin-metalloprotease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8624