Learn More
Biomatik Corporation Mouse Synaptophysin (SYP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07540
Encompass_Preferred
Mouse Synaptophysin (SYP) ELISA Kit for detecting SYP in serum, plasma, tissue homogenates, cerebrospinal fluid, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.262ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62277. Gene ID: 20977. Target Synonyms: Major synaptic vesicle protein p38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3875