Learn More
Biomatik Corporation Human Syndecan 1 (SDC1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07548
Encompass_Preferred
Human Syndecan 1 (SDC1) ELISA Kit for detecting SDC1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.66ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18827. Gene ID: 6382. Target Synonyms: CD138; SDC; SYND1; Syndecan Proteoglycan 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3877