Learn More
Biomatik Corporation Human Syntenin 2 (ST2) ELISA Kit, 96 tests
Catalog No. 501503880
Encompass_Preferred
Human Syntenin 2 (ST2) ELISA Kit for detecting ST2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H190. Gene ID: 27111. Target Synonyms: SDCBP2; SITAC18; Syndecan Binding Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3880 Supplier Biomatik Corporation Supplier No. EKU07559
Human Syntenin 2 (ST2) ELISA Kit for detecting ST2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H190. Gene ID: 27111. Target Synonyms: SDCBP2; SITAC18; Syndecan Binding Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.