Learn More
Biomatik Corporation Human Fibronectin (FN) ELISA Kit
Supplier:  Biomatik Corporation EKL57330
Encompass_Preferred
Human Fibronectin (FN) ELISA Kit for detecting FN in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.39-25ng/ml. Sensitivity: 0.17ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: P02751. Gene ID: 2335. Target Synonyms: FN1; CIG; FINC; LETS; MSF; GFND2; Anastellin; Migration-Stimulating Factor; Cold-Insoluble Globulin; Large; External; Transformation-Sensitive Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8683