Learn More
Biomatik Corporation Human Tankyrase 1 (TNKS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07569
Encompass_Preferred
Human Tankyrase 1 (TNKS1) ELISA Kit for detecting TNKS1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95271. Gene ID: 8658. Target Synonyms: TNKS; ARTD5; PARPL; PARP-5a; PARP5A; TIN1; TINF1; TNKS-1; ADP-ribosyltransferase diphtheria toxin-like 5; TRF1-Interacting Ankyrin-Related ADP-Ribose Polymerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3885