Learn More
Biomatik Corporation Human Tar DNA Binding Protein 43kDa (TDP43) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07570
Encompass_Preferred
Human Tar DNA Binding Protein 43kDa (TDP43) ELISA Kit for detecting TDP43 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4,000pg/ml. Sensitivity: 24.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13148. Gene ID: 23435. Target Synonyms: TARDBP; ALS10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3886