Learn More
Biomatik Corporation Human Target of The Antiproliferative Antibody 1 (TAPA1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07571
Encompass_Preferred
Human Target Of The Antiproliferative Antibody 1 (TAPA1) ELISA Kit for detecting TAPA1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60033. Gene ID: 975. Target Synonyms: CD81; S5.7; TAPA1; TSPAN28; Tetraspanin 28; Target Of Antiproliferative Antibody 1; 26 kDa cell surface protein TAPA-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3887