Learn More
Biomatik Corporation Human Matrix Metalloproteinase 8 (MMP8) ELISA Kit
Supplier:  Biomatik Corporation EKL59113
Encompass_Preferred
Human Matrix Metalloproteinase 8 (MMP8) ELISA Kit for detecting MMP8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 39-2500pg/ml. Sensitivity: 19pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: P22894. Gene ID: 4317. Target Synonyms: CLG2; HNC; PMNL-CL; Collagenase 2; Neutrophil Collagenase; PMNL collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8738