Learn More
Biomatik Corporation Human Tau-Protein Kinase 1 (tPK1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07575
Encompass_Preferred
Human Tau-Protein Kinase 1 (tPK1) ELISA Kit for detecting tPK1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5TCY1. Gene ID: 84630. Target Synonyms: TTBK1; BDTK; TTK; Cdk5/P20; CDK5/P23; GSK; STK31; Brain-derived tau kinase; Tau-Tubulin Kinase 1; Tau-Protein O-Hosphotransferase; Brain Protein Kinase PK40erk. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3888