Learn More
Biomatik Corporation Human Telomerase Reverse Transcriptase (TERT) ELISA Kit, 96 tests
Catalog No. 501503890
Encompass_Preferred
Human Telomerase Reverse Transcriptase (TERT) ELISA Kit for detecting TERT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14746. Gene ID: 7015. Target Synonyms: EST2; TCS1; TP2; TRT; hEST2; Telomerase catalytic subunit; Telomerase-associated protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3890 Supplier Biomatik Corporation Supplier No. EKU07587
Human Telomerase Reverse Transcriptase (TERT) ELISA Kit for detecting TERT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14746. Gene ID: 7015. Target Synonyms: EST2; TCS1; TP2; TRT; hEST2; Telomerase catalytic subunit; Telomerase-associated protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.