Learn More
Biomatik Corporation Rat Telomerase Reverse Transcriptase (TERT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07588
Encompass_Preferred
Rat Telomerase Reverse Transcriptase (TERT) ELISA Kit for detecting TERT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q673L6. Gene ID: 301965. Target Synonyms: EST2; TCS1; TP2; TRT; hEST2; Telomerase catalytic subunit; Telomerase-associated protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3891