Learn More
Biomatik Corporation Human Neuraminidase (NEU) ELISA Kit
Supplier:  Biomatik Corporation EKL59353
Encompass_Preferred
Human Neuraminidase (NEU) ELISA Kit for detecting NEU in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: Q99519. Gene ID: 4758. Target Synonyms: NEU1; SIAL1; Sialidase 1; Lysosomal Sialidase; Acetylneuraminyl hydrolase; G9 sialidase; N-acetyl-alpha-neuraminidase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8747