Learn More
Biomatik Corporation Rat Tenascin C (TNC) ELISA Kit, 96 tests
Catalog No. 501503893
Encompass_Preferred
Rat Tenascin C (TNC) ELISA Kit for detecting TNC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.131ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B2LYI9. Gene ID: 116640. Target Synonyms: TN-C; HXB; TN; GMEM; JI; Myotendinous antigen; Neuronectin; GP 150-225; Cytotactin; Hexabrachion; Glioma-associated-extracellular matrix antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3893 Supplier Biomatik Corporation Supplier No. EKU07593
Rat Tenascin C (TNC) ELISA Kit for detecting TNC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.131ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B2LYI9. Gene ID: 116640. Target Synonyms: TN-C; HXB; TN; GMEM; JI; Myotendinous antigen; Neuronectin; GP 150-225; Cytotactin; Hexabrachion; Glioma-associated-extracellular matrix antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.