Learn More
Biomatik Corporation Testosterone (Testo) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07605
Encompass_Preferred
Testosterone (Testo) ELISA Kit for detecting Testo in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 370.4-30,000pg/ml. Sensitivity: 152.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: T; Testesteron; Testeronun. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3898