Learn More
Biomatik Corporation Human Tetraspanin 1 (TSPAN1) ELISA Kit, 96 tests
Catalog No. 501503899
Encompass_Preferred
Human Tetraspanin 1 (TSPAN1) ELISA Kit for detecting TSPAN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60635. Gene ID: 10103. Target Synonyms: NET-1; Tetraspanin TM4-C; Tetraspan NET-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3899 Supplier Biomatik Corporation Supplier No. EKU07607
Human Tetraspanin 1 (TSPAN1) ELISA Kit for detecting TSPAN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60635. Gene ID: 10103. Target Synonyms: NET-1; Tetraspanin TM4-C; Tetraspan NET-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.