Learn More
Biomatik Corporation Human Tetraspanin 30 (TSPAN30) ELISA Kit, 96 tests
Catalog No. 501503900
Encompass_Preferred
Human Tetraspanin 30Cluster of Differentiation 63 (CD63) ELISA Kit for detecting CD63 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08962. Gene ID: 967. Target Synonyms: TSPAN30; LAMP-3; ME491; MLA1; OMA81H; Melanoma 1 antigen; Granulophysin; Lysosomal-associated membrane protein 3; Ocular melanoma-associated antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3900 Supplier Biomatik Corporation Supplier No. EKU07608
Human Tetraspanin 30Cluster of Differentiation 63 (CD63) ELISA Kit for detecting CD63 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08962. Gene ID: 967. Target Synonyms: TSPAN30; LAMP-3; ME491; MLA1; OMA81H; Melanoma 1 antigen; Granulophysin; Lysosomal-associated membrane protein 3; Ocular melanoma-associated antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.