Learn More
Biomatik Corporation Mouse Tetraspanin 30 (TSPAN30) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07609
Encompass_Preferred
Mouse Tetraspanin 30Cluster of Differentiation 63 (CD63) ELISA Kit for detecting CD63 in plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41731. Gene ID: 12512. Target Synonyms: TSPAN30; LAMP-3; ME491; MLA1; OMA81H; Melanoma 1 antigen; Granulophysin; Lysosomal-associated membrane protein 3; Ocular melanoma-associated antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3901