Learn More
Biomatik Corporation Human Thiopurine Methyltransferase (TPMT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07612
Encompass_Preferred
Human Thiopurine Methyltransferase (TPMT) ELISA Kit for detecting TPMT in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51580. Gene ID: 7172. Target Synonyms: Thiopurine S Methyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3902