Learn More
Biomatik Corporation Mouse Thioredoxin (Trx) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07614
Encompass_Preferred
Mouse Thioredoxin (Trx) ELISA Kit for detecting Trx in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10639. Gene ID: 22166. Target Synonyms: TXN; ADF; SASP; TRDX; TRX1; ATL-derived factor; Surface-associated sulphydryl protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3904