Learn More
Biomatik Corporation Rat Thioredoxin (Trx) ELISA Kit, 96 tests
Catalog No. 501503905
Encompass_Preferred
Rat Thioredoxin (Trx) ELISA Kit for detecting Trx in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11232. Gene ID: 116484. Target Synonyms: TXN; ADF; SASP; TRDX; TRX1; ATL-derived factor; Surface-associated sulphydryl protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3905 Supplier Biomatik Corporation Supplier No. EKU07616
Rat Thioredoxin (Trx) ELISA Kit for detecting Trx in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11232. Gene ID: 116484. Target Synonyms: TXN; ADF; SASP; TRDX; TRX1; ATL-derived factor; Surface-associated sulphydryl protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.