Learn More
Biomatik Corporation Human Toll Like Receptor 4 (TLR4) ELISA Kit
Supplier:  Biomatik Corporation EKL61084
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernatants and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: sandwich. Detection Method: sandwich. Assay Time: 3 hours. UniProt ID: O00206. Target Synonyms: CD284,TOLL,HToll. Restriction: For Research Use Only.
Catalog No. 50-228-8797