Learn More
Biomatik Corporation Human Ubiquitin Carboxyl Terminal Hydrolase L1 (UCHL1) ELISA Kit
Supplier:  Biomatik Corporation EKL61354
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernatants and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.056ng/mL. Format: 96-Well Plate. Assay Type: sandwich. Detection Method: sandwich. Assay Time: 3 hours. UniProt ID: P09936. Target Synonyms: PARK5,PGP9.5,Uch-L1,Neuron cytoplasmic protein 9.5,Ubiquitin thioesterase L1,Ubiquitin carboxyl-terminal hydrolase isozyme L1. Restriction: For Research Use Only.
Catalog No. 50-228-8804