Learn More
Biomatik Corporation Pig Thrombomodulin (TM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07631
Encompass_Preferred
Pig Thrombomodulin (TM) ELISA Kit for detecting TM in plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1SAT9. Target Synonyms: CD141; THBD; BDCA3; THRM; Fetomodulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3910