Learn More
Biomatik Corporation Rat Thrombomodulin (TM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07632
Encompass_Preferred
Rat Thrombomodulin (TM) ELISA Kit for detecting TM in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35370. Gene ID: 83580. Target Synonyms: CD141; THBD; BDCA3; THRM; Fetomodulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3911