Learn More
Biomatik Corporation Rat Thrombopoietin (TPO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07638
Encompass_Preferred
Rat Thrombopoietin (TPO) ELISA Kit for detecting TPO in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49745. Gene ID: 81811. Target Synonyms: THPO; MGDF; MKCSF; ML; MPLLG; Myeloproliferative Leukemia Virus Oncogene Ligand; Megakaryocyte Growth And Development Factor; Megakaryocyte Stimulating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3914