Learn More
Biomatik Corporation Human Thrombospondin 1 (THBS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07640
Encompass_Preferred
Human Thrombospondin 1 (THBS1) ELISA Kit for detecting THBS1 in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8,000pg/ml. Sensitivity: 61pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07996. Gene ID: 7057. Target Synonyms: TSP1; Thrombospondin-1p180. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3915