Learn More
Biomatik Corporation Mouse Aspartate Aminotransferase (AST) ELISA Kit
Supplier:  Biomatik Corporation EKL61707
Encompass_Preferred
Mouse Aspartate Aminotransferase (AST) ELISA Kit for detecting AST in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: P05201. Gene ID: 14718. Target Synonyms: cCAT; AAT; ASAT; SGOT; GOT1; Cysteine transaminase; cytoplasmic; Transaminase A; Aspartate Transaminase 1; Cytoplasmic; Glutamic-Oxaloacetic Transaminase 1; Soluble. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8820